Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mis-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129068.3 | NP_001122540.1 | 2 - 3 | 199 | 71 | |
NM_058783.8 | NP_491184.1 | 2 - 3 | 158 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |