Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058785.6 | NP_491186.2 | 5 - 6 | 1282 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |