Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: smp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058796.5 | NP_491197.3 | 6 - 7 | 1118 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |