Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: larp-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058808.5 | NP_491209.3 | 5 - 6 | 841 | 86 | |
NM_182024.8 | NP_871824.3 | 5 - 6 | 835 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |