Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hcp-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058843.8 | NP_491244.1 | 6 - 7 | 1618 | 83 | |
GenBank mRNA | AF321299.1 | - | 5 - 6 | 1585 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |