Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026325.2 | NP_001021496.1 | 7 - 8 | 1192 | 82 | |
NM_001026326.4 | NP_001021497.1 | 7 - 8 | 1080 | 82 | ||
NM_001381834.2 | NP_001367474.1 | 7 - 8 | 1080 | 82 | ||
GenBank mRNA | AF048834.1 | - | 7 - 8 | 1192 | 82 | |
AF048835.1 | - | 7 - 8 | 1192 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |