Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nab-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379751.2 | NP_001367535.1 | 2 - 3 | 215 | 103 | |
NM_001379754.2 | NP_001367538.1 | 2 - 3 | 213 | 103 | ||
NM_001382888.1 | 2 - 3 | 220 | 103 | |||
NM_001382891.1 | 1 - 2 | 123 | 103 | |||
NM_001382892.1 | 2 - 3 | 212 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |