Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndx-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058935.7 | NP_491336.2 | 2 - 3 | 429 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |