Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cogc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058938.8 | NP_491339.2 | 2 - 3 | 1219 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |