Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: met-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058939.6 | NP_491340.2 | 6 - 7 | 1530 | 108 | |
NM_182042.4 | NP_871842.2 | 6 - 7 | 1501 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |