Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356817.3 | NP_001343707.1 | 5 - 6 | 1063 | 69 | |
NM_058952.5 | NP_491353.2 | 5 - 6 | 1084 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |