Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356817.3 | NP_001343707.1 | 8 - 9 | 2038 | 75 | |
NM_058952.5 | NP_491353.2 | 8 - 9 | 2059 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |