Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001356817.3 | NP_001343707.1 | 20 - 21 | 10582 | 57 | |
NM_001356818.3 | NP_001343735.1 | 9 - 10 | 7674 | 57 | ||
NM_058952.5 | NP_491353.2 | 20 - 21 | 10603 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |