Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001330925.3 | NP_001317773.1 | 7 - 8 | 8626 | 82 | |
NM_058962.6 | NP_491363.1 | 10 - 11 | 11435 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |