Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gcy-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026429.3 | NP_001021600.1 | 10 - 11 | 2542 | 143 | |
NM_001262699.2 | NP_001249628.1 | 13 - 14 | 2436 | 143 | ||
NM_001372632.2 | NP_001360115.1 | 13 - 14 | 2428 | 143 | ||
NM_001382937.1 | 9 - 10 | 2333 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |