Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047215.5 | NP_001040680.1 | 1 - 2 | 344 | 73 | |
NM_001129040.5 | NP_001122512.1 | 1 - 2 | 346 | 73 | ||
NM_001129041.3 | NP_001122513.1 | 1 - 2 | 344 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |