Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugt-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059036.6 | NP_491437.1 | 3 - 4 | 692 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |