Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059069.4 | NP_491470.1 | 2 - 3 | 433 | 128 | |
GenBank mRNA | U14525.1 | - | 2 - 3 | 385 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |