Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059069.4 | NP_491470.1 | 7 - 8 | 1175 | 84 | |
GenBank mRNA | U14525.1 | - | 7 - 8 | 1127 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |