Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059071.7 | NP_491472.1 | 1 - 2 | 250 | 93 | |
GenBank mRNA | X98600.1 | - | 196 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |