Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: imb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059076.7 | NP_491477.2 | 13 - 14 | 2661 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |