Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dyf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059093.7 | NP_491494.2 | 8 - 9 | 940 | 72 | |
NM_182063.7 | NP_871863.1 | 6 - 7 | 721 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |