Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mes-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026321.4 | NP_001021492.1 | 14 - 15 | 2077 | 136 | |
NM_001374884.2 | NP_001361974.1 | 13 - 14 | 2024 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |