Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfe-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025876.5 | NP_001021047.1 | 2 - 3 | 852 | 95 | |
NM_001025877.6 | NP_001021048.1 | 3 - 4 | 460 | 95 | ||
NM_001025878.7 | NP_001021049.1 | 6 - 7 | 572 | 95 | ||
NM_001262849.3 | NP_001249778.1 | 6 - 7 | 555 | 95 | ||
NM_001372686.1 | NP_001359911.1 | 5 - 6 | 494 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |