Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elpc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377580.1 | NP_001364720.1 | 6 - 7 | 938 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |