Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059140.6 | NP_491541.1 | 3 - 4 | 669 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |