Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fncm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059142.4 | NP_491543.1 | 7 - 8 | 1575 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |