Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059158.7 | NP_491559.1 | 2 - 3 | 198 | 100 | |
NM_059159.4 | NP_491560.1 | 2 - 3 | 164 | 100 | ||
GenBank mRNA | AB107268.1 | - | 2 - 3 | 220 | 100 | |
AB107269.1 | - | 2 - 3 | 186 | 100 | ||
AY643538.1 | - | 2 - 3 | 188 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |