Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383080.1 | 4 - 5 | 435 | 80 | ||
NM_059174.8 | NP_491575.1 | 4 - 5 | 629 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |