Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026404.7 | NP_001021575.1 | 7 - 8 | 1064 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |