Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059229.7 | NP_491630.1 | 3 - 4 | 303 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |