Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059267.3 | NP_491668.1 | 4 - 5 | 1557 | 84 | |
GenBank mRNA | U73946.1 | - | 3 - 4 | 254 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |