Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059272.6 | NP_491673.1 | 3 - 4 | 687 | 152 | |
GenBank mRNA | AJ277649.1 | - | 3 - 4 | 595 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |