Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mtk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380377.2 | NP_001367356.1 | 13 - 14 | 4029 | 80 | |
NM_059282.5 | NP_491683.1 | 14 - 15 | 4081 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |