Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: obr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380388.1 | NP_001367357.1 | 4 - 5 | 1100 | 62 | |
NM_059290.7 | NP_491691.1 | 9 - 10 | 1937 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |