Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pgp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313481.3 | NP_001300410.1 | 13 | 3533 | 90 | |
NM_001313482.3 | NP_001300411.1 | 13 - 14 | 3632 | 90 | ||
NM_001313483.4 | NP_001300412.1 | 12 - 13 | 3636 | 90 | ||
NM_059306.6 | NP_491707.2 | 12 - 13 | 3473 | 90 | ||
GenBank mRNA | EF205592.1 | - | 12 - 13 | 3461 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |