Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025795.3 | NP_001020966.2 | 5 - 6 | 746 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |