Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383179.1 | 12 - 13 | 1219 | 76 | ||
NM_001383181.1 | 10 - 11 | 1382 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |