Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025792.4 | NP_001020963.1 | 3 - 4 | 428 | 147 | |
NM_001383179.1 | 2 - 3 | 323 | 147 | |||
NM_001383181.1 | 2 - 3 | 329 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |