Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: avr-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025792.4 | NP_001020963.1 | 7 - 8 | 816 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |