Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pmk-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171392.7 | NP_741457.2 | 1 - 2 | 116 | 71 | |
NM_171913.6 | NP_741458.2 | 1 - 2 | 105 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |