Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wsp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383137.1 | 2 - 3 | 456 | 109 | ||
NM_171393.8 | NP_741459.2 | 2 - 3 | 474 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |