Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028198.5 | NP_001023369.1 | 2 - 3 | 105 | 73 | |
NM_001307205.2 | NP_001294134.1 | 3 - 4 | 352 | 73 | ||
NM_001307206.3 | NP_001294135.1 | 2 - 3 | 105 | 73 | ||
NM_001383138.1 | 3 - 4 | 337 | 73 | |||
NM_171396.7 | NP_741462.1 | 3 - 4 | 332 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |