Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171399.6 | NP_741465.1 | 6 - 7 | 774 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |