Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spp-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171399.6 | NP_741465.1 | 7 - 8 | 841 | 94 | |
NM_171400.5 | NP_741466.1 | 5 - 6 | 2434 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |