Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: set-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060371.4 | NP_492772.1 | 1 - 2 | 227 | 108 | |
NM_182049.7 | NP_871849.1 | 1 - 2 | 92 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |