Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lam-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060374.5 | NP_492775.2 | 11 - 12 | 8834 | 121 | |
GenBank mRNA | AF074902.1 | - | 11 - 12 | 8834 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |