Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exc-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027868.5 | NP_001023039.1 | 2 - 3 | 283 | 154 | |
NM_001307099.3 | NP_001294028.1 | 1 - 2 | 82 | 154 | ||
NM_001307100.4 | NP_001294029.1 | 3 - 4 | 519 | 154 | ||
GenBank mRNA | AB060647.1 | - | 2 - 3 | 336 | 154 | |
AF339059.1 | - | 2 - 3 | 336 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |