Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028218.6 | NP_001023389.1 | 14 - 15 | 4071 | 119 | |
NM_001028219.3 | NP_001023390.1 | 15 - 16 | 4254 | 119 | ||
NM_001268536.3 | NP_001255465.1 | 10 - 11 | 3691 | 119 | ||
NM_001268537.3 | NP_001255466.1 | 9 - 10 | 3565 | 119 | ||
GenBank mRNA | AF170796.1 | - | 14 - 15 | 4060 | 119 | |
AF308447.1 | - | 14 - 15 | 4060 | 119 | ||
AF308448.1 | - | 16 - 17 | 4197 | 119 | ||
AF308449.1 | - | 11 - 12 | 3758 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |