Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pxf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028218.6 | NP_001023389.1 | 9 - 10 | 3029 | 88 | |
NM_001028219.3 | NP_001023390.1 | 9 - 10 | 3029 | 88 | ||
NM_001268536.3 | NP_001255465.1 | 5 - 6 | 2649 | 88 | ||
NM_001268537.3 | NP_001255466.1 | 4 - 5 | 2523 | 88 | ||
NM_001380435.1 | NP_001367298.1 | 5 - 6 | 2649 | 88 | ||
NM_001380436.1 | NP_001367299.1 | 4 - 5 | 2523 | 88 | ||
GenBank mRNA | AF170796.1 | - | 9 - 10 | 3018 | 88 | |
AF308447.1 | - | 9 - 10 | 3018 | 88 | ||
AF308448.1 | - | 9 - 10 | 3018 | 88 | ||
AF308449.1 | - | 6 - 7 | 2716 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |